CritiCore Cleanroom Frocks with ESD Grid
Fournisseur: VWR International
Certificats
A propos de cet article
Reusable zippered frocks are designed for controlled environments and less critical cleanroom applications. Made with durable ESD Grid Fabric, they are engineered for electrostatic dissipation with a 1% carbon grid, and also feature knit cuffs with 5% carbon Belltron fiber. Applications include micro-electronics and semi-conductor manufacturing.
- Zippered, durable, and reusable
- For micro-electronics and semi-conductor manufacturing
- All seams are laser cut or pre-serged, silicon-free thread
Garments have a mandarin collar with two snaps at top, a wide-pull zipper, and a single snap below at bottom
Available in white and navy.
Assembled with double-needle felled seams for added durability and particle migration prevention. All seams are laser-cut or pre-serged, and thread is silicon-free.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Spécifications
- Catalog No:
- 89495-854
- 89495-856
- 89495-836
- 89495-858
- 89495-838
- 89495-846
- 89495-868
- 89495-848
- 89495-870
- 89495-850
- 89495-852
- 89495-860
- 89495-840
- 89495-862
- 89495-842
- 89495-864
- 89495-844
- 89495-866
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- ESD grid fabric
- Weight:
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.
- 3.0 oz./sq. yd.