SP Bel-Art Air Operated Turbine Magnetic Stirrer, Bel-Art Products, a part of SP
# de catalogue 89049-012
Fournisseur: VWR International
Spécifications
- Description:Air-Operated Turbine Magnetic Stirrer
- Cat. no.:89049-012
- Dimensions:10.2Ø×1.9W cm (4×3/4")
Spécifications
A propos de cet article
This compact, small-footprint stirrer only uses 3–8psi (21-55kPa) air pressure and is ideal for stirring vessels up to 1 liter.
- Tubing is supplied to connect to an air supply
- All parts are made from high-impact polystyrene
- Ensured long life and easy cleanup in case of spills
The air-operated turbine magnetic stirrer can be used with magnetic stirring bars up to 2.5 cm (1") in length.
Customers Who Bought This Also Bought