Distillation Receiver, Vertical Top, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This distillation receiver is for use where separate fractions are desired without disturbing the distillation pressure
Has [ST] 14/20 joints. Receiver flasks are 5 mL. Hose connections are for use with 3/8" or 5/16" I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete [ST] 14/20 size includes the following items: vacuum adapter (89074-470), distributor (89074-460), four 5 mL flasks (89074-584), and five Dupont Delrin joint clips/clamps (89059-608). For PTFE-clad version, see 89074-462. For [ST] 24/40 version (NOT PTFE-clad), see 89055-730.
Customers Who Bought This Also Bought