About this item
A closed-loop HEPA-filtered containment glove box used for small-scale handling/weighing of active pharmaceutical ingredients ("APIs") in powder form
Glove Box also includes a closed-loop Double HEPA Filtration Package, two clear acrylic square transfer chambers (Ingress/ Egress. No gas valves), Magnehelic™ Gauge bracket with on/off switch for blower - no removable top, HEPA Capsule with manual ball valve, magnetic Interlocks on transfer chamber doors, single covered electrical receptacle, and work station ionizer, #800-AS/SPI.
The 800-AS/SPI, Anti-Static Ionizer emits alternating positive and negative ions, removing static inside the glove box. This is a non-air assisted static eliminator, (no blower/no fan). The 800-AS/SPI is perfect for working with fine powders and chemicals. 800-BIBO Containment Levels are <1.0 nanograms/cubic meter.
Features include a 3/8" thick clear acrylic construction with 3/8" thick bright white acrylic bottom, a hinged front door with oval polypropylene glove ports and door lock hasp, and white Hypalon™ gloves.