Corning® Cell and Tissue Adhesive, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Corning® Cell-Tak™ Cell and Tissue Adhesive, is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.