VWR® Heavy-Duty Ice Bags
Fournisseur: VWR International
Certificats
A propos de cet article
Made of extra strength heavy gauge Metallocene Low Density Polyethylene (LDPE) film that meets FDA/USDA specifications.
- Expandable side gussets
- Flexible poly film
This special poly film retains flexibility at low temperatures to reduce breakage. Tubular construction with a durable bottom seal, make these bags the best ice bags on the market. Side gussets expand easily to accommodate fast loading. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought