VWR® Linear Low Density Liners, Black
Supplier: VWR International
Certificates
About this item
Whereas most other LLD bags are manufactured using standard LLD materials or upgrade to a Hexene resin, these liners start with premium Octene, the highest grade of LLDPE resin available.
- Superior quality materials
- All bags over 31" long are star sealed
- Tubular construction eliminates risk of seal splitting
VWR's tubular construction eliminates the risk of side seal splitting.
Packaging: Packaged in white corrugated.
Customers Who Bought This Also Bought