VWR® Pink Anti-Static Bags
Fournisseur: VWR International
Certificats
A propos de cet article
This premium anti-static bag won't corrode polycarbonate.
- Antistatic properties
- Amine-free
- Performs in low humidity
With a surface resistivity less than 1×1012 ohms/sq VWR's 4 mm bags. This can also be heat sealed, tied, stapled or taped shut.
Certifications: Meeta the full static decay requirements of MIL-PRF-81705E, Type II and EIA-541.
Customers Who Bought This Also Bought