VWR® Linear Low Density Liner Bags
Fournisseur: VWR International
Certificats
A propos de cet article
These linear low density (LLD) Liners are a step ahead of others; whereas most other LLD bags are manufactured using standard LLD materials or upgrade to a Hexene resin, we start with premium Octene, the highest grade of LLDPE resin available.
- Durable tubular construction
- Twice as strong as LDPE bags
- All bags over 31" long are star sealed
- Manufactured with 100% renewable energy
VWR's tubular construction eliminates the risk of side seal splitting. Engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene bags.
Customers Who Bought This Also Bought