Polystyrene Test Tube, 13×75, Globe Scientific
# de catalogue 60819-970
Fournisseur: Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Diameter ×L:13 x 75
- Description:Polystyrene Test Tube
- Supplier No.:110470
Spécifications
A propos de cet article
Polystyrene tubes can withstand 1500 rpm during centrifugation.
Customers Who Bought This Also Bought