Polypropylene Test Tube, 13×75, Globe Scientific
Catalog # 60819-968
Supplier: Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Diameter ×L:13×75
- Description:Polypropylene Test Tube
- Supplier No.:110471
Specifications
About this item
Polypropylene tubes can withstand 3000 rpm.
They are autoclavable and freezable.
Frequently Bought Together