SacII, Restriction Enzymes, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
A propos de cet article
A Streptomyces lividans strain that carries the SacII gene from Streptomyces achromogenes (ATCC 12767).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.