NEB®Turbo Competent E.coli (High Effic.), New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
A propos de cet article
Chemically competent E.coli cells suitable for high efficiency transformation and rapid colony growth (6.5 hours).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.