VACUUBRAND® Vacuum Pump Oil B, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
Ensure the reliability of your VACUUBRAND® Rotary Vane or Chemistry-HYBRID vacuum pump by using Pump Oil B
- Optimized for VACUUBRAND rotary vane and Chemistry-HYBRID pumps
- Low vapor pressure, even at high temperatures
- Excellent stability for Chemistry applications
- 1- and 5-liter containers
Pump Oil B has excellent viscosity characteristics, good chemical resistance, low vapor pressure even at high temperatures, and better stability when pumping oxidants such as acid and alkaline vapors. Superior to standard mineral oils. Suitable for all VACUUBRAND® RE-, RZ- and RC-series pumps.
Customers Who Bought This Also Bought