CritiCore Indura Women's Flame Resistant Lab Coats
Fournisseur: VWR International
Certificats
A propos de cet article
CritiCore has developed these flame-resistant lab coats specifically to fit the female form, while also providing the protection required in many of today's high-tech laboratories and research facilities.
- Princess-belt on back
- Flame-resistant identification patches sewn on the back and both sleeves for easy recognition
- Quick-release snap front for fast removal
- Safety-sleeve cuff with tunneled elastic
- Two oversized waist pockets and one breast pocket with a convenient pencil slot
- Vent openings on the sides allow access to pant pockets
These coats are made with 100% cotton fabric, durable enough to withstand industrial laundering while maintaining fire-resistant properties for the life of the garment.
Certifications: CAT I
Ordering information: Available only in blue. Coats in sizes X-Small to XX-Large are stocked. For sizes 3X-Large to 6X-Large, extended lead time may be required. Contact a Avantor sales representative for more information.
Customers Who Bought This Also Bought