MICROFLEX® NeoTouch™ 25-101, Chemical-Resistant Gloves, Neoprene, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
This glove is made with neoprene, an alternative to natural rubber latex that protects against Type I allergies.
Ideal applications include assembly and inspection, lab analysis and technical work, and pharmaceutical manufacturing - especially when resistance to acids is required. These gloves have a roll-beaded cuff, and are powder-free.
- Resistant to acids, bases and alcohols
- Roll-beaded cuff, powder free
This glove is made with neoprene, an alternative to natural rubber latex that protects against Type I allergies.
Ideal applications include assembly and inspection, lab analysis and technical work, and pharmaceutical manufacturing - especially when resistance to acids is required.
Their exclusive formulation provides superior comfort.
Certifications: This is an exam- and food-grade glove and complies with FDA food handling requirements (21 CFR 177.2600).
Spécifications
- Catalog No:
- 89011-910
- 89011-908
- 89011-902
- 89011-904
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Finish:
- Textured Fingertips
- Textured Fingertips
- Textured Fingertips
- Textured Fingertips