Drying Tube, 75°, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Angled gooseneck-type drying tube features a dessicant area that is 140 mm in length.
Customers Who Bought This Also Bought
Angled gooseneck-type drying tube features a dessicant area that is 140 mm in length.