Drying Tube Adapter, Straight, Glassco
# de catalogue 89426-424
Fournisseur: Glassco
Spécifications
- Description:Drying Tube Adapter, Straight
- Cat. no.:89426-424
- ST Joint Size:19/22
- Supplier No.:041.470.02
Spécifications
A propos de cet article
Drying tube adapters are used for keeping a reaction vessel free of moisture.
- Straight
- Top [ST] joint
- Bottom [ST] joint
Adapter is equipped with standard taper joints at top and bottom.
Customers Who Bought This Also Bought