Allihn Condenser, Glassco
# de catalogue 89426-686
Fournisseur: Glassco
Spécifications
- Length:400 mm
- Description:Allihn Condenser
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:190.471.06
Spécifications
A propos de cet article
Condensers feature standard taper inner joint at bottom and reinforced standard taper outer joint at top.
Customers Who Bought This Also Bought