Chromatography Column, Heavy Wall Design, Glassco
Supplier: Glassco
Certificates
Certificates
About this item
General purpose column features a PTFE stopcock and a top outer joint for flash chromatography.
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
238,72 $
WHEATON® MicroLiter Septa for Screw-Thread Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
53,82 $
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
1 294,78 $ to 1 294,78 $
WHEATON® MicroLiter 20 mm Bi-Metal Crimp Seals, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
30,99 $
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
104,01 $