Claisen Adapter, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Claisen adapters provide dual entry into a variety of flasks.
- Top [ST] joint
- Bottom [ST] joint
Manufactured from borosilicate 3.3 glass.
Customers Who Bought This Also Bought