Distillation Adapter, Vigreux, Glassco
# de catalogue 89426-370
Fournisseur: Glassco
Spécifications
- Description:Distillation Adapter, Vigreux
- Cat. no.:89426-370
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 10/18
- Supplier No.:028.470.03A
Spécifications
A propos de cet article
This distillation adapter is designed to accommodate an immersion thermometer.
- Vigreux-type
- Accommodates thermometer
Adapter accommodates a 76mm immersion thermometer.
Customers Who Bought This Also Bought