Distillation Adapter, 105°, with Inner Drip Joint, Glassco
# de catalogue 89426-354
Fournisseur: Glassco
Spécifications
- Description:Distillation Adapter, 105°, with Inner Drip Joint
- Cat. no.:89426-354
- Bottom [ST] Joint Size:14/20
- Top ST Joint Size:[ST] 14/20
- Supplier No.:025.475.01
Spécifications
A propos de cet article
Adapter is equipped with an inner drip joint at the bottom and an outer joint at the top.
- 105° angle bend
- Top outer joint
- Bottom inner drip joint
Fabricated from borosilicate 3.3 glass.
Customers Who Bought This Also Bought