Distillation Trap Adapter, Glassco
# de catalogue 89426-382
Fournisseur: Glassco
Spécifications
- Capacity:100 ml
- Description:Distillation Trap Adapter
- Cat. no.:89426-382
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:030.476.04A
Spécifications
A propos de cet article
This trap adapter is placed above the flask to prevent carry-over of reactions which have a tendency to foam.
- Inner vapor tube
- Manufactured from borosilicate 3.3 glass
Adapter has a capacity of 100mL.
Customers Who Bought This Also Bought