Distillation Adapter, 105°, Glassco
# de catalogue 89426-374
Fournisseur: Glassco
Spécifications
- Description:Distillation Adapter, 105°
- Cat. no.:89426-374
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:025.470.06
Spécifications
A propos de cet article
This Distillation Adapter is also designed for use as a rotary evaporator washer.
- 105° angle bend
- Top [ST] joint
- Bottom [ST] joint
Also designed for use as rotary evaporator washer, this Distillation Adapter is very simple and handy. When filled with ethanol or acetone, a sudden finger release will spray the mashing solvent into the Rotavapor Condenser, thus cleaning the entire system.
Customers Who Bought This Also Bought