Inlet Adapter, with Hose Connection, Glassco
# de catalogue 89426-402
Fournisseur: Glassco
Spécifications
- Description:Inlet Adapter, with Hose Connection
- Cat. no.:89426-402
- ST Joint Size:24/40
- Supplier No.:022.471.03
Spécifications
A propos de cet article
This inlet adapter is equipped with a vaccum take-off and drip tip.
- Hose connection
- Drip tip
- Manufactured from borosilicate 3.3 glass
Customers Who Bought This Also Bought