Flushing Adapter, Glassco
# de catalogue 89426-436
Fournisseur: Glassco
Spécifications
- Description:Flushing Adapter
- ST Joint Size:24/40
- Supplier No.:048.202.01
Spécifications
A propos de cet article
The flushing adapter is used to flush and fill attached vessels.
- T-bore PTFE stopcock
- [ST] joint
Adapter is equipped with a 2mm T-bore PTFE stopcock.
Customers Who Bought This Also Bought