Vacuum Adapter, with PTFE Stopcock, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Vacuum adapters are designed to regulate the flow of gases or liquids into reaction systems.
- PTFE stopcock
- Hose connection
Stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration with standard inner taper.
Customers Who Bought This Also Bought