Vacuum Adapter, with 1:5 Fluoropolymer Resin Stopcock, Glassco
# de catalogue 89426-406
Fournisseur: Glassco
Spécifications
- Description:Vacuum Adapter, with 1:5 Fluoropolymer Resin Stopcock
- Cat. no.:89426-406
- ST Joint Size:19/22
- Supplier No.:034.470.02
Spécifications
A propos de cet article
This vacuum adapter features a hose connection and stopcock.
- Hose connection
- Fluoropolymer resin stopcock
- [ST] joint
Adapter is equipped with an 8mm O.D. hose connection and 2mm fluoropolymer resin stopcock.
Customers Who Bought This Also Bought