Inlet Adapter, Fritted, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
The fritted disc of this inlet adapter prevents solids from being introduced into the system.
- Extra coarse disc
- Serrated hose connection
- Inner joint
Adapters are equipped with an extra coarse disc between the serrated hose connection and the inner joint that prevents solids from being introduced into the system. Hose connection has a O.D. of 8mm.
Customers Who Bought This Also Bought