Inlet Adapter, 90°, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Adapters feature an inner joint and a 90° hose connection.
- Inner [ST] joint
- 90° angle hose connection
- Manufactured from borosilicate 3.3 glass
Use to adapt a hose connection.
Customers Who Bought This Also Bought