Vaccum/Argon Adapter (Introgen), Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Adapters feature a straight hose connection at top and male joint.
- Top hose connection
- Male [ST] joint
- Manufactured from borosilicate 3.3 glass.
Use adapter for vacuum or argon applications.
Customers Who Bought This Also Bought