Inlet or Thermometer Adapter, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Adapters have a thread at the top for use with a compression cap and O-Ring.
- Top thread accommodates compression cap or O-Ring
- Can be used as syringe port
To use as a syringe port, simply replace the O-Ring with a fluoropolymer resin-lined septum.
Customers Who Bought This Also Bought