Corning® 384-Well, Clear Polystyrene Microplates, Corning
Supplier: Corning
Certificates
About this item
PS, flat bottom.
- Total well volume of 112 μl; working well volume of 20 to 80 μl
- Cell culture plates sterilized by gamma radiation and certified non pyrogenic
- Universal optics NBS™ plate manufactured using an advanced polymer with high clarity and improved chemical resistance
- Lids supplied with sterile plates; other lids can be ordered separately
White walled plates enhance luminescent signals and have low background luminescence and fluorescence. Black walled plates have low background fluorescence, minimal light scatter.
Specifications
- Catalog No:
- 77776-798
- 29444-092
- 29444-096
- 29444-094
- 89092-712
- 29445-115
- CA89089-780
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk