Bushing Adapter, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Bushing adapters allow for compact assemblies of dissimilar joints.
- Top [ST] joint
- Bottom [ST] joint
- Manufactured from borosilicate 3.3 glass
Bushing-style adapters have upper and lower standard taper joints.
Customers Who Bought This Also Bought