Round Bottom Flask, Double Neck, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Round bottom flask with same sized joints
Heavy wall design. Manufactured from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform wall thickness.
Customers Who Bought This Also Bought