Round Bottom Flask, Single Neck, Glassco
Fournisseur: Glassco
Certificats
A propos de cet article
Heavy wall design with single neck
Made from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform thickness.
Customers Who Bought This Also Bought