PTFE Covered Stainless Steel Stir Shaft with Retreat Curve Agitator, 10 mm, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This PTFE covered stainless steel shaft features a solid PTFE retreat curve agitator
Featuring a retreat angle of 30 degrees and, for a better grip by the stir motor chuck, 50mm of exposed stainless steel shaft.
Customers Who Bought This Also Bought