A propos de cet article
The Boekel Scientific family of Water Baths offers a variety of sizes to meet all of your needs and is ideal for general laboratory applications
Equipped with an adjustable over temperature safety cutout which sounds an audible alarm and shuts the heater off helps protect your samples while the recessed control panel helps protect against spills. Also featured is a seamless passivated tank to prevent against rust and corrosion.
The space saving design allows for the most efficient use of your bench space and the Microprocessor Controller makes for easy temperature selection, rapid heat-up and excellent stability. The special boiling mode is a convenient feature for melting agar as well as other applications that require a boiling temperature (to utilize the Boiling Mode, you will need to purchase the Stainless Steel Gabled Lid accessory).
Spécifications
- Catalog No:
- 97004-338
- 97004-344
- 97004-342
- Temperature range:
- Ambient +5°C to 100°C
- Ambient +5°C to 100°C
- Ambient +5°C to 100°C
- Timer:
- Settable to 99 hrs, 59 min or "OFF"
- Settable to 99 hrs, 59 min or "OFF"
- Settable to 99 hrs, 59 min or "OFF"