Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Schlenk filter tubes feature an integral glass filter disc at the tube's mid-line and two sidearms with either a glass stopcock or a 0-3 inlet valve
Paired top and bottom inner joints. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought