Gas Washing Bottle with Ace Thred, Ace Glass Incorporated
Catalog # 89428-056
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:500 ml (16.9 oz.)
- Description:Gas Washing Bottle with Ace Thred
- Cat. no.:89428-056
- Supplier No.:7161-10
Specifications
About this item
Newly designed gas washing bottle with the convenience of Ace-Threds.
- Bottom fritted disc is Porosity B, 70 to 100 µm
Center washing tube fits into borosilicate glass bottle via #25 Ace-Thred with nylon busing and FETFE O-Ring. Washing tube has #7 Ace-Safe inlet/outlet ports with ¼" hose connections.
Customers Who Bought This Also Bought