ContactUsLinkComponent
Lab Armor® Bead Baths, 6 L, Shel Lab, Sheldon
Lab Armor® Bead Baths, 6 L, Shel Lab, Sheldon
Fournisseur:  Sheldon Manufacturing
Certificats
Lab Armor® Bead Baths, 6 L, Shel Lab, Sheldon
Lab Armor® Bead Baths, 6 L, Shel Lab, Sheldon
Fournisseur:  Sheldon Manufacturing
Certificats
Spécifications
  • Catalog No:
  • 89409-218
  • 89409-216
  • Dimensions:
  • 15.2 x 30.5 x 15.2 cm (6 x 12 x 6"), Tub
  • 15.2 x 30.5 x 15.2 cm (6 x 12 x 6"), Tub
  • Exterior dimensions:
  • 21.6 x 40.4 x 31.2 cm (8.5 x 15.9 x 12.3")
  • 21.6 x 40.4 x 31.2 cm (8.5 x 15.9 x 12.3")