Platinum-Cured Silicone Pump Tubing on a Spool, NewAge Industries
Fournisseur: NewAge Industries
Certificats
A propos de cet article
Extremely flexible tubing is specially formulated to withstand repeated compression and release for consistent, reliable performance in peristaltic pumps.
- Sterilizable by autoclave and gamma radiation
- Imparts no taste or odor to critical stream
Low-volatile grade, platinum-cured silicone tubing is produced in a certified ISO Class 6 (FED-STD-209E Class 1000/M4.5) cleanroom for use in critical pharmaceutical, biomedical, laboratory, food, beverage, and fragrance applications. Designed to maintain a long service life, this tubing features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
Tubing has undergone extensive physical, chemical, and biological testing and meets USP Class VI and FDA CFR 177.2600 standards; compound meets ISO 10993 standards.
Certifications: Certified free of animal-derived components.
Ordering information: Supplied in (100' to 500') lengths. Sealed in polyethylene bags to ensure cleanliness. Quality control and lot traceability identification on bags and boxes. Complete validation package available upon request.
Caution: Note: Not intended for implantation, continuous stream applications, or elevated pressure levels.
Customers Who Bought This Also Bought