VWR® CryoPro® Plastic Storage Boxes
Fournisseur: VWR International
Certificats
A propos de cet article
Polycarbonate construction for use in mechanical and LN2 freezers.
- Grid format for lids
- Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 25, 81, and 100 cell grids to assist with inventory control.
Customers Who Bought This Also Bought