VWR® Angled Thermometer Storage Trays
Catalog # 89203-047
Supplier: VWR International
Specifications
- Description:Angled Thermometer Storage Tray, 14 Slots
- Dimensions:442L x 340W x 30D mm (17.4 x 13.4 x 1.2"), Tray
- Cat. no.:89203-047
Specifications
About this item
Recyclable trays feature a built-in angle to prevent separation of thermometer liquid.
- UV coated
- Made from 2mm polystyrene
3- or 14-slot versions available with numbered slots.
Frequently Bought Together