A propos de cet article
SHEL LAB high-performance water baths are accurate, easy to use, safe and durable
This durable and dependable water bath comes with a variety of safety and convenience features. It has a unique lid design that allows condensation to drain back into the tank without spilling onto the work area. We have also incorporated a gasket around the water bath tank to prevent water from dripping onto the controls. A warm air jacketed design radiates heat to the sides and bottom of the tank, eliminating hot spots. The unit has a non-contact, recessed heating element that will prevent burnouts if the water level is too low.
The water bath tank is constructed of stainless steel and does not have seams that could leak. The radius corners of the tank make cleaning easy. The unit's housings have a high quality textured powder coat finish. This electrostatically applied finish not only eliminates rust and chipping but is resistant to nicks, scratches and corrosives. The SHEL LAB constant temperature digital water baths are perfect for conducting a host of applications including; bacteriological examinations, food processing/QC procedures and microbiology assays just to name a few. (Also available in 220V).
The digital set/digital read P.I.D. temperature controller maximizes rapid heat-up without temperature overshoot. A microprocessor achieves precise temperature control regardless of how the unit is loaded. Calibration is performed directly from the front panel. This is a convenient feature for facilities concerned with IQ/OQ/PQ validation.
Spécifications
- Catalog No:
- 89409-384
- 89409-380
- Temperature range:
- 5°C above Ambient to 80°C
- 5°C above Ambient to 80°C
- Temperature uniformity:
- +/- 0.2°C at 37°C
- +/- 0.2°C at 37°C