HiTrap Columns, Streptavidin HP, Cytiva
Fournisseur: Cytiva
Certificats
A propos de cet article
HiTrap Streptavidin HP is a column prepacked with Streptavidin Sepharose High Performance for purification of biotinylated proteins and biomolecules.
- Convenient, fast, reliable purification of biotinylated compounds
- The strong interactions of biotin and streptavidin, as well as the somewhat weaker interactions of 2-iminobiotin and streptavidin are utilised
- The column can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System
Streptavidin Sepharose™ High Performance is also available in lab packs, using the same purified streptavidin isolated from Streptomyces avidinii bound to Sepharose™ High Performance. The prepacked medium ensures reproducibility, high performance, and excellent physical and chemical stability.
Spécifications
- Catalog No:
- CA95055-892L
- 95055-892
- Description:
- CA95055-892L
- 95055-892
- Max. pressure:
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- Storage:
- 4 to 30 °C, 20% Ethanol, 0.1 M Potassium Phosphate buffer, pH 8
- 4 to 30 °C, 20% Ethanol, 0.1 M Potassium Phosphate buffer, pH 8