Collection Plates for 96-Well Plates, Cytiva
# de catalogue CA95056-282L
Fournisseur: Cytiva
Spécifications
- Well Volume:<500 µL
- Description:Collection Plates
- Cat. no.:CA95056-282L
- Supplier No.:28-4039-43
Spécifications
A propos de cet article
Made from biocompatible PP. Collection plates, 96-well, designed for collection of eluted samples in parallel from screening plates such as MultiTrap™ and PreDictor™ plates.
- Convenient collection of samples in 96-well format
- Excellent fit to prepacked MultiTrap plates and PreDictor plates
- V-bottom for minimal loss of sample
- Collection of < 500µL volumes of sample per well
Customers Who Bought This Also Bought