Non-Sparking Shovel, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Made of durable, corrosion-resistant polypropylene
- Hardwood-core handle resists hundreds of pounds of pressure
- Comes in two pieces for easy storage
Excellent for use with flammable, explosive or volatile chemicals
Customers Who Bought This Also Bought