Rotary Evaporator Connecting Tubes, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Connecting glass tubes for the receiver assembly are available in either poly-coated or plain, uncoated formats
DN25 inner ball joints have CAPFE (PTFE-encapsulated silicone rubber) O-Rings, size –217.
Delivery: Replacement borosilicate glass tubes for Buchi® R-220, R-220EX, and R-220SE rotary evaporators. The PTFE valve stem (89235-604) includes three Kalrz O-Rings.
Customers Who Bought This Also Bought